This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01022957_m1
See other ADGRV1 GE Assays ›
Gene Symbol:
ADGRV1
Gene Name:
adhesion G protein-coupled receptor V1
Gene Aliases:
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
Chromosome Location:
Chr.5: 90558681 - 91164226 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01022957_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRV1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032119.3 NP_115495.3 87 - 88 18529 102
NR_003149.1 87 - 88 18446 102
XM_017009963.1 XP_016865452.1 87 - 88 18669 102
XM_017009964.1 XP_016865453.1 87 - 88 18666 102
XM_017009965.1 XP_016865454.1 87 - 88 18551 102
XM_017009966.1 XP_016865455.1 86 - 87 18588 102
XM_017009967.1 XP_016865456.1 86 - 87 18458 102
XM_017009968.1 XP_016865457.1 86 - 87 18489 102
XM_017009972.1 XP_016865461.1 59 - 60 11951 102
XM_017009973.1 XP_016865462.1 64 - 65 13162 102
GenBank mRNA AB014586.2 - 14 - 15 3115 102
AF055084.1 - 23 - 24 5700 102
AF435925.1 - 87 - 88 18532 102
AK024416.1 - 17 - 18 3905 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500008055.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000442.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRV1

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
nervous system development
visual perception
sensory perception of sound
single organismal cell-cell adhesion
photoreceptor cell maintenance
maintenance of organ identity
neurological system process
sensory perception of light stimulus
G-protein coupled receptor activity
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05006332.hg.1 ENST00000296619.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05006332.hg.1 ENST00000399043.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05006332.hg.1 ENST00000405460.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05006332.hg.1 ENST00000425867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05006332.hg.1 NM_032119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05006332.hg.1 NR_003149.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05006332.hg.1 TC05000442.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 ENST00000405460.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 ENST00000425867.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 NM_032119.3 Refseq
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 NR_003149.1 Refseq
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 ENST00000405460.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 ENST00000425867.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 NM_032119.3 Refseq
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 NR_003149.1 Refseq
Affymetrix Human Clariom D Assay PSR0500155258.hg.1 TC0500008055.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0