This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01022902_m1
See other ADGRV1 GE Assays ›
Gene Symbol:
ADGRV1
Gene Name:
adhesion G protein-coupled receptor V1
Gene Aliases:
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
Chromosome Location:
Chr.5: 90558681 - 91164226 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01022902_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRV1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032119.3 NP_115495.3 27 - 28 5764 73
NR_003149.1 27 - 28 5764 73
XM_017009963.1 XP_016865452.1 27 - 28 5883 73
XM_017009964.1 XP_016865453.1 27 - 28 5880 73
XM_017009965.1 XP_016865454.1 27 - 28 5765 73
XM_017009966.1 XP_016865455.1 26 - 27 5802 73
XM_017009967.1 XP_016865456.1 26 - 27 5672 73
XM_017009968.1 XP_016865457.1 27 - 28 5883 73
XM_017009969.1 XP_016865458.1 27 - 28 5883 73
XM_017009970.1 XP_016865459.1 27 - 28 5883 73
XM_017009971.1 XP_016865460.1 27 - 28 5883 73
XM_017009973.1 XP_016865462.1 4 - 5 480 73
XM_017009974.1 XP_016865463.1 27 - 28 5883 73
GenBank mRNA AF435925.1 - 27 - 28 5764 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500008055.hg
This assay targets the same location as HTA transcript cluster(s) TC05000442.hg

Additional Information:

This assay is in the 5'UTR region of the XM_017009973.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRV1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
nervous system development
visual perception
sensory perception of sound
single organismal cell-cell adhesion
photoreceptor cell maintenance
maintenance of organ identity
neurological system process
sensory perception of light stimulus
G-protein coupled receptor activity
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05006258.hg.1 ENST00000296619.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05006258.hg.1 ENST00000399043.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05006258.hg.1 ENST00000405460.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05006258.hg.1 NM_032119.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05006258.hg.1 NR_003149.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05006258.hg.1 TC05000442.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500155188.hg.1 ENST00000405460.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500155188.hg.1 NM_032119.3 Refseq
Affymetrix Human Clariom D Assay PSR0500155188.hg.1 NR_003149.1 Refseq
Affymetrix Human Clariom D Assay PSR0500155188.hg.1 ENST00000405460.6 Ensembl
Affymetrix Human Clariom D Assay PSR0500155188.hg.1 NM_032119.3 Refseq
Affymetrix Human Clariom D Assay PSR0500155188.hg.1 NR_003149.1 Refseq
Affymetrix Human Clariom D Assay PSR0500155188.hg.1 TC0500008055.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0