This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01022364_m1
See other GARS GE Assays ›
Gene Symbol:
GARS
Gene Name:
glycyl-tRNA synthetase
Gene Aliases:
CMT2D, DSMAV, GlyRS, HMN5, SMAD1
Chromosome Location:
Chr.7: 30594735 - 30634033 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01022364_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GARS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001316772.1 NP_001303701.1 6 - 7 921 154
NM_002047.3 NP_002038.2 6 - 7 763 154
XM_006715686.2 XP_006715749.1 7 - 8 923 154
GenBank mRNA AK074524.1 - 6 - 7 824 154
AK295490.1 - 7 - 8 783 154
AK310351.1 - 6 - 7 772 154
BC007722.2 - 6 - 7 781 4299853 154
BC007755.2 - 6 - 7 739 4109682 154
D30658.1 - 6 - 7 758 154
U09510.1 - 6 - 7 936 154
U09587.1 - 6 - 7 728 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700007098.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000198.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GARS

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase translational protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
glycyl-tRNA aminoacylation
diadenosine tetraphosphate biosynthetic process
bis(5'-nucleosyl)-tetraphosphatase (asymmetrical) activity
glycine-tRNA ligase activity
ATP binding
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000389266.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000444666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000444666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000444666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000444666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000444666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000444666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000444666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000454308.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000465748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000465748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000465748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000465748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000465748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000465748.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000470392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000470392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000470392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000470392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000478124.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000484093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000484093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000484093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000484093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000484093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000485784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000485784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000485784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000485784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000485784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000485784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000496643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000496643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000496643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000496643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000496643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000496643.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000627489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000627489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 ENST00000627489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007098.hg.1 NM_002047.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0