Assay Details
Gene Symbol: GLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR749593.1 | - | 1 - 2 | 1800 | 161 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000320717.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000338435.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409215.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000409626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000412247.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000417154.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000457316.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000461965.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000468352.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000469774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000471443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000479552.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000495444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000496170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | ENST00000629173.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_001256310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010275.hg.1 | NM_014905.4 | Refseq |
