This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01020669_g1
See other SHKBP1 GE Assays ›
Gene Symbol:
SHKBP1
Gene Name:
SH3KBP1 binding protein 1
Gene Aliases:
PP203, Sb1
Chromosome Location:
Chr.19: 40576851 - 40591399 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01020669_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SHKBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138392.3 NP_612401.2 4 - 5 307 62
XM_006723474.1 XP_006723537.1 4 - 5 307 62
XM_006723475.1 XP_006723538.1 4 - 5 307 62
XM_011527492.1 XP_011525794.1 4 - 5 307 62
XM_017027473.1 XP_016882962.1 4 - 5 307 62
GenBank mRNA AF258553.1 - 4 - 5 306 62
AK075057.1 - 4 - 5 303 62
AK300819.1 - 4 - 5 303 62
AK312748.1 - 4 - 5 271 62
BC022855.1 - 4 - 5 278 4102901 62
DB038927.1 - 4 - 5 307 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK300819.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SHKBP1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

positive regulation of epidermal growth factor receptor signaling pathway
protein homooligomerization

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19007733.hg.1 ENST00000291842.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19007733.hg.1 ENST00000446701.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19007733.hg.1 NM_138392.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19007733.hg.1 TC19000564.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0