Assay Details
Gene Symbol: EIF5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015904.3 | NP_056988.3 | 14 - 15 | 2445 | 223 | |
GenBank mRNA | AB018284.1 | - | 14 - 15 | 2411 | 223 | |
AF078035.1 | - | 14 - 15 | 2378 | 223 | ||
AJ006776.1 | - | 14 - 15 | 2401 | 223 | ||
AK001386.1 | - | 5 - 6 | 683 | 223 | ||
AL133563.1 | - | 11 - 12 | 1997 | 223 | ||
BC032639.1 | - | 14 - 15 | 2367 | 5496504 | 223 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000603.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02009551.hg.1 | ENST00000289371.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009551.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02009551.hg.1 | TC02000603.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160016.hg.1 | TC0200008587.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
