This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01016802_m1
See other EZH2 GE Assays ›
Gene Symbol:
EZH2
Gene Name:
enhancer of zeste 2 polycomb repressive complex 2 subunit
Gene Aliases:
ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
Chromosome Location:
Chr.7: 148807372 - 148884662 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01016802_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EZH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004456.4 NP_004447.2 8 - 9 1102 109
XM_011515883.2 XP_011514185.1 9 - 10 1077 109
XM_011515884.2 XP_011514186.1 8 - 9 1209 109
XM_011515885.2 XP_011514187.1 8 - 9 3518 109
XM_011515886.2 XP_011514188.1 9 - 10 1226 109
XM_011515887.2 XP_011514189.1 8 - 9 1075 109
XM_011515888.2 XP_011514190.1 8 - 9 1197 109
XM_011515889.2 XP_011514191.1 8 - 9 999 109
XM_011515890.2 XP_011514192.1 7 - 8 3428 109
XM_011515891.2 XP_011514193.1 6 - 7 1162 109
XM_011515892.2 XP_011514194.1 8 - 9 3546 109
XM_011515894.2 XP_011514196.1 7 - 8 927 109
XM_011515895.2 XP_011514197.1 7 - 8 3401 109
XM_011515897.2 XP_011514199.1 9 - 10 1253 109
XM_011515898.2 XP_011514200.1 9 - 10 1180 109
XM_011515899.2 XP_011514201.1 8 - 9 3548 109
XM_011515901.2 XP_011514203.1 8 - 9 3548 109
XM_017011817.1 XP_016867306.1 8 - 9 3544 109
XM_017011820.1 XP_016867309.1 7 - 8 958 109
GenBank mRNA BC010858.2 - 8 - 9 1031 3901250 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700012959.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001993.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EZH2

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA methylation
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of epithelial to mesenchymal transition
regulation of gliogenesis
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
cerebellar cortex development
hippocampus development
response to estradiol
negative regulation of transcription elongation from RNA polymerase II promoter
regulation of cell proliferation
regulation of circadian rhythm
positive regulation of MAP kinase activity
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of GTPase activity
negative regulation of epidermal cell differentiation
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
negative regulation of retinoic acid receptor signaling pathway
rhythmic process
negative regulation of striated muscle cell differentiation
cellular response to hydrogen peroxide
G1 to G0 transition
histone H3-K27 methylation
protein localization to chromatin
positive regulation of protein serine/threonine kinase activity
positive regulation of dendrite development
negative regulation of G1/S transition of mitotic cell cycle
core promoter binding
DNA binding
chromatin binding
RNA binding
protein binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
chromatin DNA binding
histone methyltransferase activity
ribonucleoprotein complex binding
sequence-specific DNA binding
histone methyltransferase activity (H3-K27 specific)
promoter-specific chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000320356.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000350995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000460911.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000476773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000478654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000483967.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000536783.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 ENST00000541220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 NM_001203247.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 NM_001203248.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 NM_001203249.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 NM_004456.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 NM_152998.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030622.hg.1 TC07001993.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000320356.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000350995.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000460911.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000476773.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000478654.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000483012.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000483967.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000492143.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000498186.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_001203247.1 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_001203248.1 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_001203249.1 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_004456.4 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_152998.2 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000320356.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000350995.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000460911.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000476773.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000478654.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000483012.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000483967.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000492143.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 ENST00000498186.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_001203247.1 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_001203248.1 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_001203249.1 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_004456.4 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 NM_152998.2 Refseq
Affymetrix Human Clariom D Assay PSR0700193449.hg.1 TC0700012959.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0