This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01016162_g1
See other SMC3 GE Assays ›
Gene Symbol:
SMC3
Gene Name:
structural maintenance of chromosomes 3
Gene Aliases:
BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1
Chromosome Location:
Chr.10: 110567691 - 110604634 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01016162_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005445.3 NP_005436.1 16 - 17 1795 77
GenBank mRNA AF020043.1 - 16 - 17 1760 77
AF067163.1 - 4 - 5 443 77
AK289771.1 - 16 - 17 1791 77
AK315845.1 - 16 - 17 1794 77
BC047324.1 - 16 - 17 1690 5493263 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008896.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000802.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMC3

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA replication
DNA repair
mitotic spindle organization
sister chromatid cohesion
mitotic sister chromatid cohesion
mitotic nuclear division
meiotic nuclear division
signal transduction
protein sumoylation
stem cell population maintenance
negative regulation of DNA endoreduplication
cell division
chromatin binding
microtubule motor activity
protein binding
ATP binding
mediator complex binding
dynein binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10009612.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009612.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009612.hg.1 TC10000802.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay PSR1000160816.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000160816.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay PSR1000160816.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000160816.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay PSR1000160816.hg.1 TC1000008896.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0