This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01016159_m1
See other SMC3 GE Assays ›
Gene Symbol:
SMC3
Gene Name:
structural maintenance of chromosomes 3
Gene Aliases:
BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1
Chromosome Location:
Chr.10: 110567691 - 110604634 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01016159_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005445.3 NP_005436.1 13 - 14 1436 101
GenBank mRNA AF020043.1 - 13 - 14 1401 101
AF067163.1 - 1 - 2 83 101
AK289771.1 - 13 - 14 1432 101
AK315845.1 - 13 - 14 1435 101
BC047324.1 - 13 - 14 1331 5493263 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008896.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000802.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF067163.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMC3

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA replication
DNA repair
mitotic spindle organization
sister chromatid cohesion
mitotic sister chromatid cohesion
mitotic nuclear division
meiotic nuclear division
signal transduction
protein sumoylation
stem cell population maintenance
negative regulation of DNA endoreduplication
cell division
chromatin binding
microtubule motor activity
protein binding
ATP binding
mediator complex binding
dynein binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009609.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009609.hg.1 NM_005445.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009609.hg.1 TC10000802.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000160813.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000160813.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay PSR1000160813.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000160813.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay PSR1000160813.hg.1 TC1000008896.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000361804.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 ENST00000462899.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq
Affymetrix Human Clariom D Assay TC1000008896.hg.1 NM_005445.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0