Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010988.hg.1 |
ENST00000358776.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010988.hg.1 |
ENST00000368869.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010988.hg.1 |
ENST00000496730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010988.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010988.hg.1 |
TC10000896.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
ENST00000541070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009169.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
ENST00000358776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
ENST00000368869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
ENST00000411816.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
NM_001609.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162938.hg.1 |
TC1000009169.hg.1 |
Clariom D TC |