This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01014023_m1
See other GLS GE Assays ›
Gene Symbol:
GLS
Gene Name:
glutaminase
Gene Aliases:
AAD20, GAC, GAM, GLS1, KGA
Chromosome Location:
Chr.2: 190880821 - 190965552 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01014023_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256310.1 NP_001243239.1 4 - 5 991 122
NM_014905.4 NP_055720.3 4 - 5 991 122
XM_005246467.3 XP_005246524.1 4 - 5 991 122
XM_006712435.1 XP_006712498.1 4 - 5 991 122
XM_017003829.1 XP_016859318.1 4 - 5 991 122
GenBank mRNA AB020645.1 - 4 - 5 977 122
AF097492.1 - 4 - 5 625 122
AF158555.1 - 5 - 6 971 122
AF223943.1 - 4 - 5 746 122
AF279697.1 - 1 - 2 76 122
AF327434.1 - 4 - 5 957 122
AK289747.1 - 5 - 6 967 122
BC038507.1 - 5 - 6 967 5263220 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200010275.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001122.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF279697.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLS

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

suckling behavior
regulation of respiratory gaseous exchange by neurological system process
glutamate biosynthetic process
glutamine catabolic process
chemical synaptic transmission
cellular amino acid biosynthetic process
glutamate secretion
protein homotetramerization
glutaminase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000320717.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000338435.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409428.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000409626.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000412247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000412247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000412247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000412247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000412247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000412247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000417154.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000457316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000461965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000468352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000468352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000468352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000468352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000468352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000468352.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000469774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000469774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000469774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000469774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000469774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000469774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000471443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000471443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000471443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000471443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000471443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000479552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000495444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000495444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000495444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000495444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000495444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000496170.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000496170.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000496170.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000496170.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000496170.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 ENST00000629173.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_001256310.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010275.hg.1 NM_014905.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header