Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021527.hg.1 |
ENST00000179765.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021527.hg.1 |
ENST00000379441.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021527.hg.1 |
ENST00000549884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021527.hg.1 |
ENST00000551812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021527.hg.1 |
NM_013449.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021527.hg.1 |
TC12001608.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008222.hg.1 |
ENST00000605342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008222.hg.1 |
ENST00000605342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008222.hg.1 |
ENST00000605342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008222.hg.1 |
ENST00000605342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000379441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000549787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000549884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000551812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
NM_001300905.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
NM_013449.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000379441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000549787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000549884.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
ENST00000551812.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
NM_001300905.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
NM_013449.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184441.hg.1 |
TC1200010926.hg.1 |
Clariom D TC |