This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01011552_g1
See other HRC GE Assays ›
Gene Symbol:
HRC
Gene Name:
histidine rich calcium binding protein
Gene Aliases:
-
Chromosome Location:
Chr.19: 49151198 - 49155443 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01011552_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HRC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002152.2 NP_002143.1 1 - 2 2017 100
GenBank mRNA BC069795.1 - 1 - 2 2000 7262494 100
BC069802.1 - 1 - 2 2000 7262506 100
BC094691.1 - 1 - 2 2017 30529366 100
BC112355.1 - 1 - 2 1982 40034092 100
M60052.1 - 1 - 2 2000 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900011127.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001709.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HRC

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
muscle contraction
positive regulation of heart rate
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of peptidyl-serine phosphorylation
positive regulation of heart contraction
regulation of cytosolic calcium ion concentration
negative regulation of cytosolic calcium ion concentration
regulation of ryanodine-sensitive calcium-release channel activity
regulation of cell communication by electrical coupling involved in cardiac conduction
positive regulation of relaxation of cardiac muscle
regulation of calcium ion transmembrane transport
calcium ion binding
protein binding
ion channel binding
ATPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023354.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023354.hg.1 ENST00000434964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023354.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023354.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023355.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023355.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023355.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023350.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023350.hg.1 ENST00000434964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023350.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023350.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023349.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023349.hg.1 ENST00000391863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023349.hg.1 ENST00000434964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023349.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023349.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023345.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023345.hg.1 ENST00000391863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023345.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023345.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023352.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023352.hg.1 ENST00000434964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023352.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023352.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023357.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023357.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023357.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023348.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023348.hg.1 ENST00000391863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023348.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023348.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023356.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023356.hg.1 ENST00000434964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023356.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023356.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023346.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023346.hg.1 ENST00000391863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023346.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023346.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19023347.hg.1 ENST00000252825.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19023347.hg.1 NM_002152.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19023347.hg.1 TC19001709.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193127.hg.1 TC1900011127.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900193132.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193132.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193132.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193132.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193132.hg.1 TC1900011127.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900193130.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193130.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193130.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193130.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193130.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193130.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193130.hg.1 TC1900011127.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595167.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay TC1900011127.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 ENST00000252825.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 ENST00000595625.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 ENST00000598858.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 NM_002152.2 Refseq
Affymetrix Human Clariom D Assay PSR1900193129.hg.1 TC1900011127.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0