This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01011515_m1
See other IL12B GE Assays ›
Gene Symbol:
IL12B
Gene Name:
interleukin 12B
Gene Aliases:
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
Chromosome Location:
Chr.5: 159314783 - 159330473 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01011515_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL12B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002187.2 NP_002178.2 2 - 3 131 123
GenBank mRNA AF180563.1 - 1 - 2 95 123
BC067498.1 - 1 - 2 93 6971868 123
BC067499.1 - 1 - 2 93 6971869 123
BC067500.1 - 1 - 2 93 6971872 123
BC067501.1 - 1 - 2 93 6971873 123
BC067502.1 - 1 - 2 93 6971874 123
BC074723.2 - 2 - 3 120 30915173 123
EF173865.1 - 1 - 2 57 123
M65272.1 - 2 - 3 131 123
M65290.1 - 1 - 2 102 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012646.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001997.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL12B

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
natural killer cell activation involved in immune response
positive regulation of T-helper 1 type immune response
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
negative regulation of inflammatory response to antigenic stimulus
cell cycle arrest
response to UV-B
positive regulation of activation of JAK2 kinase activity
cell migration
cytokine-mediated signaling pathway
sensory perception of pain
sexual reproduction
natural killer cell activation
negative regulation of interleukin-10 production
negative regulation of interleukin-17 production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-17 production
positive regulation of tumor necrosis factor production
positive regulation of natural killer cell activation
positive regulation of natural killer cell proliferation
positive regulation of mononuclear cell proliferation
positive regulation of tissue remodeling
positive regulation of smooth muscle cell apoptotic process
regulation of cytokine biosynthetic process
T-helper 1 type immune response
T-helper cell differentiation
interferon-gamma biosynthetic process
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
positive regulation of NF-kappaB import into nucleus
regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat4 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
defense response to protozoan
positive regulation of memory T cell differentiation
negative regulation of growth of symbiont in host
positive regulation of interferon-gamma biosynthetic process
positive regulation of osteoclast differentiation
positive regulation of cell adhesion
negative regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
positive regulation of inflammatory response
defense response to Gram-negative bacterium
positive regulation of NK T cell activation
positive regulation of NK T cell proliferation
defense response to virus
cellular response to lipopolysaccharide
cellular response to interferon-gamma
positive regulation of T-helper 17 type immune response
positive regulation of T-helper 17 cell lineage commitment
cytokine receptor activity
cytokine activity
interleukin-12 receptor binding
protein binding
growth factor activity
interleukin-12 alpha subunit binding
identical protein binding
protein homodimerization activity
interleukin-23 receptor binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027816.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027816.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027816.hg.1 TC05001997.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05027817.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027817.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027817.hg.1 TC05001997.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05027819.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027819.hg.1 NM_002187.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027819.hg.1 TC05001997.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0500188222.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500188222.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay PSR0500188222.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500188222.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay PSR0500188222.hg.1 TC0500012646.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay TC0500012646.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay PSR0500188219.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500188219.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay PSR0500188219.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500188219.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay PSR0500188219.hg.1 TC0500012646.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500188220.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500188220.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay PSR0500188220.hg.1 ENST00000231228.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500188220.hg.1 NM_002187.2 Refseq
Affymetrix Human Clariom D Assay PSR0500188220.hg.1 TC0500012646.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0