This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01010707_m1
See other GPR180 GE Assays ›
Gene Symbol:
GPR180
Gene Name:
G protein-coupled receptor 180
Gene Aliases:
ITR
Chromosome Location:
Chr.13: 94601850 - 94634645 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01010707_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR180
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_180989.5 NP_851320.1 2 - 3 389 98
GenBank mRNA AK126794.1 - 2 - 3 294 98
AK289850.1 - 2 - 3 636 98
BC033906.1 - 2 - 3 364 98
BC052243.1 - 2 - 3 336 6083073 98
CD110068.1 - 2 - 3 416 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300007731.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000330.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK126794.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR180

Gene Ontology Categories:

Function(s)

G-protein coupled receptor signaling pathway
response to pheromone

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13003450.hg.1 ENST00000376958.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13003450.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13003450.hg.1 TC13000330.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300152133.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300152133.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay PSR1300152133.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300152133.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay PSR1300152133.hg.1 TC1300007731.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0