Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014481.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014481.hg.1 |
ENST00000529051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014481.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014481.hg.1 |
TC11001150.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000524471.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
ENST00000530061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009407.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170526.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170526.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170526.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170526.hg.1 |
ENST00000344762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170526.hg.1 |
ENST00000529051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170526.hg.1 |
NM_025004.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170526.hg.1 |
TC1100009407.hg.1 |
Clariom D TC |