This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01009761_g1
See other CCDC15 GE Assays ›
Gene Symbol:
CCDC15
Gene Name:
coiled-coil domain containing 15
Gene Aliases:
-
Chromosome Location:
Chr.11: 124954121 - 125041489 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01009761_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCDC15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025004.2 NP_079280.2 1 - 2 247 78
XM_017018346.1 XP_016873835.1 1 - 2 247 78
XM_017018347.1 XP_016873836.1 1 - 2 247 78
XM_017018348.1 XP_016873837.1 1 - 2 247 78
XM_017018350.1 XP_016873839.1 1 - 2 247 78
XM_017018351.1 XP_016873840.1 1 - 2 247 78
XM_017018353.1 XP_016873842.1 1 - 2 247 78
GenBank mRNA AK023277.1 - 1 - 2 247 78
BC018540.1 - 1 - 2 161 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100009407.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001150.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK023277.1,BC018540.1,NM_025004.2,XM_017018346.1,XM_017018347.1,XM_017018348.1,XM_017018350.1,XM_017018351.1,XM_017018353.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CCDC15

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11014477.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014477.hg.1 ENST00000529051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014477.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11014477.hg.1 TC11001150.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11014476.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014476.hg.1 ENST00000529051.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11014476.hg.1 NM_025004.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11014476.hg.1 TC11001150.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000524471.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 ENST00000530061.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay TC1100009407.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay PSR1100170520.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170520.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170520.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay PSR1100170520.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170520.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170520.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay PSR1100170520.hg.1 TC1100009407.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100170518.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170518.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170518.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay PSR1100170518.hg.1 ENST00000344762.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170518.hg.1 ENST00000529051.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100170518.hg.1 NM_025004.2 Refseq
Affymetrix Human Clariom D Assay PSR1100170518.hg.1 TC1100009407.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0