Assay Details
Gene Symbol: STMN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015894.3 | NP_056978.2 | 1 - 2 | 288 | 104 | |
NR_075070.1 | 1 - 2 | 288 | 104 | |||
GenBank mRNA | AK074860.1 | - | 1 - 2 | 100 | 104 | |
AK094112.1 | - | 1 - 2 | 723 | 104 | ||
AK310010.1 | - | 1 - 2 | 123 | 104 | ||
BC009381.2 | - | 1 - 2 | 145 | 4124704 | 104 | |
BC025234.1 | - | 1 - 2 | 145 | 4894236 | 104 | |
DA161621.1 | - | 1 - 2 | 288 | 104 | ||
DA181040.1 | - | 1 - 2 | 272 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20001037.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK074860.1,AK094112.1,BC009381.2,BC025234.1,NM_015894.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
microtubule depolymerization
nervous system development
regulation of microtubule polymerization or depolymerization
cytoplasmic microtubule organization
neuron projection development
negative regulation of Rac protein signal transduction
regulation of GTPase activity
regulation of cytoskeleton organization
tubulin binding
protein domain specific binding
microtubule depolymerization
nervous system development
regulation of microtubule polymerization or depolymerization
cytoplasmic microtubule organization
neuron projection development
negative regulation of Rac protein signal transduction
regulation of GTPase activity
regulation of cytoskeleton organization
tubulin binding
protein domain specific binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20014217.hg.1 | ENST00000370053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20014217.hg.1 | ENST00000540534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20014217.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20014217.hg.1 | TC20001037.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20014216.hg.1 | ENST00000370053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20014216.hg.1 | ENST00000540534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20014216.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20014216.hg.1 | TC20001037.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20014219.hg.1 | ENST00000370053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20014219.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20014219.hg.1 | TC20001037.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000168620.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168620.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168620.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168620.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168620.hg.1 | TC2000009823.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000168619.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168619.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168619.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168619.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168619.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168619.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168619.hg.1 | TC2000009823.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000168621.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168621.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168621.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168621.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168621.hg.1 | TC2000009823.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009823.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000370053.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000540534.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000631920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000632676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | ENST00000634126.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | NM_001276310.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | NM_015894.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | NR_075070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000168614.hg.1 | TC2000009823.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 145191 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 148186 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 177832 | - | - |
