This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01007988_m1
See other ALDH2 GE Assays ›
Gene Symbol:
ALDH2
Gene Name:
aldehyde dehydrogenase 2 family (mitochondrial)
Gene Aliases:
ALDH-E2, ALDHI, ALDM
Chromosome Location:
Chr.12: 111766887 - 111809985 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01007988_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000690.3 NP_000681.2 11 - 12 103
NM_001204889.1 NP_001191818.1 10 - 11 103
GenBank mRNA AK223373.1 - 11 - 12 103
AK301375.1 - 10 - 11 103
AK314856.1 - 11 - 12 103
AY621070.1 - 11 - 12 103
BC002967.1 - 11 - 12 3543343 103
BC071839.1 - 11 - 12 5477768 103
CB851683.1 - 2 - 3 572 103
CR456991.1 - 11 - 12 103
K03001.1 - 8 - 9 103
M26760.1 - 8 - 9 103
X05409.1 - 11 - 12 103
Y00109.1 - 11 - 12 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200012707.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12003212.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALDH2

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
alcohol metabolic process
ethanol catabolic process
ethanol oxidation
aldehyde dehydrogenase (NAD) activity
aldehyde dehydrogenase [NAD(P)+] activity
electron carrier activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000551450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 ENST00000551450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12011487.hg.1 ENST00000261733.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12011487.hg.1 ENST00000416293.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12011487.hg.1 ENST00000552234.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12011487.hg.1 ENST00000553044.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12011487.hg.1 NM_000690.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12011487.hg.1 NM_001204889.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12011487.hg.1 TC12003212.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000551450.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 ENST00000551450.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay TC1200012707.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000261733.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000416293.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000548536.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 ENST00000549106.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 NM_000690.3 Refseq
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 NM_001204889.1 Refseq
Affymetrix Human Clariom D Assay PSR1200201653.hg.1 TC1200012707.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0