Assay Details
Gene Symbol: GRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005160.3 | NP_005151.2 | 6 - 7 | 650 | 135 | |
XM_011529975.2 | XP_011528277.1 | 5 - 6 | 360 | 135 | ||
GenBank mRNA | AK058103.1 | - | 5 - 6 | 440 | 135 | |
AK292234.1 | - | 6 - 7 | 516 | 135 | ||
AK310398.1 | - | 6 - 7 | 518 | 135 | ||
BC036797.1 | - | 7 - 8 | 594 | 5590378 | 135 | |
BX387339.2 | - | 6 - 7 | 695 | 135 | ||
X69117.1 | - | 6 - 7 | 503 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000165.hg
Additional Information:
This assay is in the 5'UTR region of the XM_011529975.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22003095.hg.1 | ENST00000324198.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003095.hg.1 | ENST00000545323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003095.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22003095.hg.1 | TC22000165.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | ENST00000324198.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | ENST00000455558.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | ENST00000619906.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | NM_005160.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200148164.hg.1 | TC2200006935.hg.1 | Clariom D TC |
