This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01007265_m1
See other GRK3 GE Assays ›
Gene Symbol:
GRK3
Gene Name:
G protein-coupled receptor kinase 3
Gene Aliases:
ADRBK2, BARK2
Chromosome Location:
Chr.22: 25564894 - 25729294 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01007265_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRK3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005160.3 NP_005151.2 3 - 4 405 176
GenBank mRNA AK292234.1 - 3 - 4 271 176
AK310398.1 - 3 - 4 273 176
BC036797.1 - 4 - 5 349 5590378 176
BX387339.2 - 3 - 4 450 176
X69117.1 - 3 - 4 258 176

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200006935.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000165.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GRK3

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
signal transduction
receptor internalization
protein kinase activity
G-protein coupled receptor kinase activity
ATP binding
beta-adrenergic receptor kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000418510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000418510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000418510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22003092.hg.1 ENST00000324198.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22003092.hg.1 ENST00000545323.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22003092.hg.1 NM_005160.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22003092.hg.1 TC22000165.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay PSR2200148161.hg.1 TC2200006935.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000324198.10 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000418510.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000418510.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000418510.1 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000455558.2 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 ENST00000619906.4 Ensembl
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq
Affymetrix Human Clariom D Assay TC2200006935.hg.1 NM_005160.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0