This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01006841_g1
See other SEMA4C GE Assays ›
Gene Symbol:
SEMA4C
Gene Name:
semaphorin 4C
Gene Aliases:
M-SEMA-F, SEMACL1, SEMAF, SEMAI
Chromosome Location:
Chr.2: 96859736 - 96870943 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01006841_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEMA4C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017789.4 NP_060259.4 7 - 8 792 88
XM_006712606.3 XP_006712669.2 7 - 8 896 88
XM_011511378.2 XP_011509680.1 7 - 8 900 88
XM_011511379.2 XP_011509681.1 7 - 8 897 88
XM_011511380.1 XP_011509682.1 7 - 8 1235 88
XM_011511381.1 XP_011509683.1 7 - 8 869 88
XM_011511382.2 XP_011509684.1 7 - 8 828 88
XM_011511383.1 XP_011509685.1 7 - 8 840 88
XM_017004393.1 XP_016859882.1 6 - 7 1322 88
XM_017004394.1 XP_016859883.1 5 - 6 444 88
GenBank mRNA AB051526.1 - 5 - 6 1022 88
AY358842.1 - 7 - 8 765 88
BC017476.1 - 1 - 2 68 88
BC109103.1 - 7 - 8 721 40035252 88
BC109104.1 - 7 - 8 718 40035253 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200013541.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002113.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SEMA4C

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
neural tube closure
cell migration in hindbrain
cerebellum development
positive regulation of cell migration
positive regulation of stress-activated MAPK cascade
muscle cell differentiation
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
protein binding
semaphorin receptor binding
neuropilin binding
chemorepellent activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02034284.hg.1 ENST00000305476.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02034284.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02034284.hg.1 TC02002113.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02034285.hg.1 ENST00000305476.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02034285.hg.1 NM_017789.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02034285.hg.1 TC02002113.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200199151.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200199151.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200199151.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay PSR0200199151.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200199151.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200199151.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay PSR0200199151.hg.1 TC0200013541.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000442264.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000449330.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000467747.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000474420.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay TC0200013541.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay PSR0200199150.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200199150.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200199150.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay PSR0200199150.hg.1 ENST00000305476.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200199150.hg.1 ENST00000482925.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200199150.hg.1 NM_017789.4 Refseq
Affymetrix Human Clariom D Assay PSR0200199150.hg.1 TC0200013541.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0