Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034276.hg.1 |
ENST00000305476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034276.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034276.hg.1 |
TC02002113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012118.hg.1 |
ENST00000566971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000442264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000449330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013541.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000305476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000467747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000474420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
ENST00000482925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
NM_017789.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199142.hg.1 |
TC0200013541.hg.1 |
Clariom D TC |