This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01006519_m1
See other IQSEC3 GE Assays ›
Gene Symbol:
IQSEC3
Gene Name:
IQ motif and Sec7 domain 3
Gene Aliases:
-
Chromosome Location:
Chr.12: 66794 - 178460 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01006519_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IQSEC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170738.1 NP_001164209.1 4 - 5 1988 55
NM_015232.1 NP_056047.1 3 - 4 1491 55
XM_011520958.2 XP_011519260.1 4 - 5 2076 55
XM_011520959.2 XP_011519261.1 4 - 5 2076 55
XM_011520960.1 XP_011519262.1 4 - 5 2075 55
XM_011520961.1 XP_011519263.1 3 - 4 1491 55
XM_017019310.1 XP_016874799.1 4 - 5 2076 55
XM_017019311.1 XP_016874800.1 4 - 5 2076 55
GenBank mRNA AB029033.1 - 1 - 2 663 55
AK091953.1 - 3 - 4 1491 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200006438.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000003.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IQSEC3

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

actin cytoskeleton organization
regulation of ARF protein signal transduction
positive regulation of GTPase activity
ARF guanyl-nucleotide exchange factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12000036.hg.1 ENST00000326261.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000036.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000036.hg.1 ENST00000538872.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000036.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000036.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000036.hg.1 TC12000003.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12000032.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000032.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000032.hg.1 TC12000003.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000544511.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 ENST00000544511.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000037.hg.1 ENST00000326261.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000037.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000037.hg.1 ENST00000538872.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12000037.hg.1 NM_001170738.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000037.hg.1 NM_015232.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12000037.hg.1 TC12000003.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144062.hg.1 TC1200006438.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144063.hg.1 TC1200006438.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200144057.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200144057.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144057.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1200144057.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay PSR1200144057.hg.1 TC1200006438.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000382841.2 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000537151.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000538872.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000540907.5 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000544511.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 ENST00000544511.1 Ensembl
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_001170738.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq
Affymetrix Human Clariom D Assay TC1200006438.hg.1 NM_015232.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0