Assay Details
Gene Symbol: RBM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032120.3 | NP_115496.2 | 5 | 3526 | 123 | |
GenBank mRNA | BC007365.2 | - | 1 | 844 | 123 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | peroxisomal biogenesis factor 1 | PEX1 |
HMLR1, PBD1A, PBD1B, ZWS, ZWS1 |
-8.3kb | Anti-sense | Chr. 7 - 92487023 - 92528531 on NCBI Build GRCh38 |
Downstream | family with sequence similarity 133 member B | FAM133B | 20.3kb | Anti-sense | Chr. 7 - 92560758 - 92590394 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864641 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864642 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05613517 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864637 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864638 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864643 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05613519 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 440203 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P59714 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P59714 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P4036 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 21491 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 29501 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P112322 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P112322 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4075 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P59714 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P59714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15988919 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864608 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864616 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613496 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864620 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613496 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864620 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864616 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15988919 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864608 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P4036 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P59714 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P112322 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P59714 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P4036 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 30913 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 11742 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01754976 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988919 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988921 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613510 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613456 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613518 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613507 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613512 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988901 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613521 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613500 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613470 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613505 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613498 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613524 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613523 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988902 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613459 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613515 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613497 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613503 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988898 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613462 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988903 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613492 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613464 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613527 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613466 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613469 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613495 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613516 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1553881_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011781.hg.1 | ENST00000613371.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1553881_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553881_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at5 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034709 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034709 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 145063 | - | - |
