This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01003107_m1
See other BAZ2B GE Assays ›
Gene Symbol:
BAZ2B
Gene Name:
bromodomain adjacent to zinc finger domain 2B
Gene Aliases:
WALp4
Chromosome Location:
Chr.2: 159315312 - 159712439 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01003107_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BAZ2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110586.1 1 - 2 120 99
XM_006712461.3 XP_006712524.2 1 - 2 95 99
XM_017003918.1 XP_016859407.1 1 - 2 124 99
XM_017003919.1 XP_016859408.1 1 - 2 115 99
XM_017003920.1 XP_016859409.1 1 - 2 88 99
GenBank mRNA BX641037.1 - 1 - 2 109 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BX641037.1, XM_006712461.3, XM_017003918.1, XM_017003919.1, XM_017003920.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay PSR0200207104.hg.1 TC0200014725.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000418770.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000453016.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay TC0200014725.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay PSR0200207102.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207102.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay PSR0200207102.hg.1 ENST00000607836.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200207102.hg.1 NR_110586.1 Refseq
Affymetrix Human Clariom D Assay PSR0200207102.hg.1 TC0200014725.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0