This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01000705_m1
See other TAF8 GE Assays ›
Gene Symbol:
TAF8
Gene Name:
TATA-box binding protein associated factor 8
Gene Aliases:
II, TAF, TAFII-43, TAFII43, TBN
Chromosome Location:
Chr.6: 42050513 - 42083274 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01000705_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAF8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138572.2 NP_612639.2 4 - 5 395 131
XM_005248849.2 XP_005248906.1 4 - 5 384 131
XM_011514296.1 XP_011512598.1 4 - 5 386 131
XM_017010241.1 XP_016865730.1 4 - 5 386 131
XM_017010242.1 XP_016865731.1 4 - 5 549 131
XM_017010243.1 XP_016865732.1 4 - 5 549 131
XM_017010244.1 XP_016865733.1 4 - 5 384 131
XM_017010245.1 XP_016865734.1 4 - 5 384 131
GenBank mRNA AF465841.1 - 4 - 5 369 131
AK057383.1 - 4 - 5 386 131
AK303097.1 - 4 - 5 553 131
AL833401.1 - 4 - 5 385 131
BC119678.1 - 4 - 5 386 40116287 131
BC119679.1 - 4 - 5 386 40116290 131
BI828714.1 - 4 - 5 393 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008035.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000579.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAF8

Gene Ontology Categories:

Function(s) Process(es)

inner cell mass cell proliferation
cell differentiation
snRNA transcription from RNA polymerase II promoter
regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
maintenance of protein location in nucleus
protein binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 ENST00000372977.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 ENST00000372978.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 ENST00000372982.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 ENST00000456846.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 ENST00000465926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 ENST00000494547.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06006513.hg.1 TC06000579.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000486070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000486070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000486070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay PSR0600155699.hg.1 TC0600008035.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372977.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372978.7 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000372982.8 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000456846.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000465926.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000472818.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482432.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000482926.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000486070.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000486070.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000486070.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 ENST00000494547.5 Ensembl
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq
Affymetrix Human Clariom D Assay TC0600008035.hg.1 NM_138572.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0