Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
ENST00000252773.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
ENST00000428389.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
ENST00000519716.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
ENST00000550896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
ENST00000551649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
ENST00000552293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
NM_001080421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018088.hg.1 |
TC19001278.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000252773.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000519716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000550896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000551649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000552293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000601528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
NM_001080421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000252773.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000519716.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000550896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000551649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000552293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
ENST00000601528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
NM_001080421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181917.hg.1 |
TC1900009984.hg.1 |
Clariom D TC |