Assay Details
Gene Symbol: CREB3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052854.3 | NP_443086.1 | 6 - 7 | 1382 | 133 | |
XM_006718380.3 | XP_006718443.1 | 6 - 7 | 1359 | 133 | ||
GenBank mRNA | AB063321.1 | - | 6 - 7 | 1003 | 133 | |
AK075538.1 | - | 1 - 2 | 153 | 133 | ||
AK314072.1 | - | 6 - 7 | 1360 | 133 | ||
BC014097.1 | - | 6 - 7 | 1025 | 4546950 | 133 | |
BC015781.1 | - | 6 - 7 | 1011 | 4856946 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000394.hg
Additional Information:
This assay is in the 5'UTR region of the AK075538.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
transcription from RNA polymerase II promoter
multicellular organism development
endoplasmic reticulum unfolded protein response
extracellular matrix constituent secretion
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
protein binding
cAMP response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding
osteoblast differentiation
transcription from RNA polymerase II promoter
multicellular organism development
endoplasmic reticulum unfolded protein response
extracellular matrix constituent secretion
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
protein binding
cAMP response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11004885.hg.1 | ENST00000288400.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004885.hg.1 | ENST00000446415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004885.hg.1 | ENST00000529193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004885.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11004885.hg.1 | TC11000394.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000534787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000534787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000534787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11004886.hg.1 | ENST00000288400.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004886.hg.1 | ENST00000446415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004886.hg.1 | ENST00000529193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004886.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11004886.hg.1 | TC11000394.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100152680.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152680.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152680.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152680.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152680.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100152680.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100152680.hg.1 | TC1100007445.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000527342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000530518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000534616.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000534787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000534787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000534787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000616094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | ENST00000621158.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007445.hg.1 | NM_052854.3 | Refseq |
