This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00999400_m1
See other CRTAM GE Assays ›
Gene Symbol:
CRTAM
Gene Name:
cytotoxic and regulatory T-cell molecule
Gene Aliases:
CD355
Chromosome Location:
Chr.11: 122838419 - 122872643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00999400_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRTAM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019604.3 NP_062550.2 6 - 7 777 154
XM_011542900.2 XP_011541202.1 5 - 6 703 154
GenBank mRNA AF001622.1 - 6 - 7 728 154
AK313578.1 - 6 - 7 775 154
BC070266.1 - 6 - 7 777 30418882 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100009334.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CRTAM

Gene Ontology Categories:

Function(s) Process(es)

T cell mediated cytotoxicity
detection of tumor cell
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of cytokine secretion
regulation of immune response
activated T cell proliferation
detection of stimulus
receptor activity
receptor binding
protein homodimerization activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay PSR1100170064.hg.1 TC1100009334.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000227348.8 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533416.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 ENST00000533709.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_001304782.1 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq
Affymetrix Human Clariom D Assay TC1100009334.hg.1 NM_019604.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0