Assay Details
Gene Symbol: CABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300896.1 | NP_001287825.1 | 1 - 2 | 232 | 73 | |
GenBank mRNA | BC033167.1 | - | 1 - 2 | 232 | 4549347 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC033167.1, NM_001300896.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159936.hg.1 | TC1100008138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100159948.hg.1 | TC1100008138.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000325656.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000438189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000538060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542025.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000542233.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545040.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000545777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | ENST00000602912.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_001300896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008138.hg.1 | NM_145200.3 | Refseq |
