This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00998386_g1
See other SRMS GE Assays ›
Gene Symbol:
SRMS
Gene Name:
src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
Gene Aliases:
C20orf148, PTK70, SRM, dJ697K14.1
Chromosome Location:
Chr.20: 63539924 - 63547504 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00998386_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRMS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080823.3 NP_543013.1 4 - 5 825 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000009817.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20001034.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SRMS

Gene Ontology Categories:

Function(s) Process(es)

transmembrane receptor protein tyrosine kinase signaling pathway
negative regulation of signal transduction
cell migration
cell differentiation
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20014150.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20014150.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20014150.hg.1 TC20001034.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR20014151.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20014151.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20014151.hg.1 TC20001034.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay TC2000009817.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay PSR2000168538.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000168538.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay PSR2000168538.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000168538.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay PSR2000168538.hg.1 TC2000009817.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000168537.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000168537.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay PSR2000168537.hg.1 ENST00000217188.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000168537.hg.1 NM_080823.3 Refseq
Affymetrix Human Clariom D Assay PSR2000168537.hg.1 TC2000009817.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0