This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00998001_g1
See other MAP4K5 GE Assays ›
Gene Symbol:
MAP4K5
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 5
Gene Aliases:
GCKR, KHS, KHS1, MAPKKKK5
Chromosome Location:
Chr.14: 50418493 - 50561126 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00998001_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAP4K5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006575.4 NP_006566.2 29 - 30 2549 128
NM_198794.2 NP_942089.1 29 - 30 2597 128
XM_006720013.3 XP_006720076.1 29 - 30 2444 128
XM_011536377.2 XP_011534679.1 28 - 29 2455 128
XM_011536378.2 XP_011534680.1 29 - 30 2294 128
XM_011536379.2 XP_011534681.1 25 - 26 2405 128
XM_011536380.2 XP_011534682.1 16 - 17 3191 128
GenBank mRNA AK124738.1 - 15 - 16 2982 128
AK313612.1 - 28 - 29 2355 128
BC036013.1 - 29 - 30 2487 5272190 128
U77129.1 - 29 - 30 2549 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400009131.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001120.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MAP4K5

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKKK activity
protein phosphorylation
activation of JUN kinase activity
intracellular signal transduction
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
MAP kinase kinase kinase kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000013125.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000364703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000554990.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000555216.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557124.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557124.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557124.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557124.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557210.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 ENST00000557578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_006575.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009131.hg.1 NM_198794.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0