This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00997904_m1
See other CAMTA1 GE Assays ›
Gene Symbol:
CAMTA1
Gene Name:
calmodulin binding transcription activator 1
Gene Aliases:
CANPMR
Chromosome Location:
Chr.1: 6785324 - 7769706 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00997904_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAMTA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015215.3 NP_056030.1 21 - 22 5166 141
XM_011541084.2 XP_011539386.1 22 - 23 5051 141
XM_011541090.2 XP_011539392.1 22 - 23 4712 141
XM_017000775.1 XP_016856264.1 20 - 21 4960 141
XM_017000778.1 XP_016856267.1 21 - 22 4691 141
XM_017000779.1 XP_016856268.1 21 - 22 4642 141
GenBank mRNA AB020640.3 - 21 - 22 5166 141
BC151835.1 - 21 - 22 5166 8860115 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100006707.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000096.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CAMTA1

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
neuromuscular process controlling balance
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000303635.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000461580.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467404.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467404.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467404.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000467404.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000470648.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000472283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000472283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000472283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000473578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000473578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000473578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000473578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000473578.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476163.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000476864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000482934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000482934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000482934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000482934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000482934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000482934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000482934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000486138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000486138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000486138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000486138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490738.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490738.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490738.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490738.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000490905.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000495233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000557126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000557126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000557126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000557126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000557126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 ENST00000557126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001195563.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001195563.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001195563.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001195563.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001195563.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001195563.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_001242701.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NM_015215.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006707.hg.1 NR_038934.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0