This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00997375_m1
See other NUB1 GE Assays ›
Gene Symbol:
NUB1
Gene Name:
negative regulator of ubiquitin like proteins 1
Gene Aliases:
BS4, NUB1L, NYREN18
Chromosome Location:
Chr.7: 151341699 - 151378461 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00997375_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243351.1 NP_001230280.1 10 - 11 1187 120
NM_016118.4 NP_057202.3 10 - 11 1187 120
XM_005250011.2 XP_005250068.1 10 - 11 1252 120
XM_017012304.1 XP_016867793.1 10 - 11 1246 120
XM_017012305.1 XP_016867794.1 10 - 11 1246 120
XM_017012306.1 XP_016867795.1 10 - 11 1268 120
XM_017012307.1 XP_016867796.1 10 - 11 1249 120
XM_017012308.1 XP_016867797.1 10 - 11 1246 120
XM_017012309.1 XP_016867798.1 10 - 11 1252 120
XM_017012310.1 XP_016867799.1 11 - 12 1349 120
XM_017012311.1 XP_016867800.1 11 - 12 1253 120
XM_017012312.1 XP_016867801.1 11 - 12 1222 120
XM_017012313.1 XP_016867802.1 11 - 12 1349 120
XM_017012314.1 XP_016867803.1 11 - 12 1225 120
GenBank mRNA AF300717.1 - 10 - 11 1176 120
AF459743.1 - 10 - 11 1135 120
AY129295.1 - 10 - 11 1135 120
BC046354.2 - 9 - 10 1103 6070128 120
BG105733.1 - 3 - 4 419 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700009700.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001016.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NUB1

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
response to interferon-gamma
response to tumor necrosis factor
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000413040.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000460712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000460712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000460712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000460712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000460712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000460712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000468404.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470229.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470229.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470229.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470229.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470229.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470229.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470229.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000470316.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000477666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000477666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000477666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000477666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000477666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000477666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000477666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480714.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000480907.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000483358.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000493588.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000497987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000497987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000497987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000497987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000497987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000497987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000497987.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 ENST00000568733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_001243351.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009700.hg.1 NM_016118.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0