This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00994075_m1
See other RNPS1 GE Assays ›
Gene Symbol:
RNPS1
Gene Name:
RNA binding protein with serine rich domain 1
Gene Aliases:
E5.1
Chromosome Location:
Chr.16: 2253116 - 2268412 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00994075_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RNPS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286625.1 NP_001273554.1 4 - 5 698 77
NM_001286626.1 NP_001273555.1 3 - 4 705 77
NM_001286627.1 NP_001273556.1 3 - 4 705 77
NM_006711.4 NP_006702.1 4 - 5 759 77
NM_080594.3 NP_542161.1 4 - 5 893 77
NR_104485.1 3 - 4 827 77
XM_005255048.1 XP_005255105.1 4 - 5 986 77
XM_005255049.3 XP_005255106.1 4 - 5 2451 77
XM_017022874.1 XP_016878363.1 4 - 5 1352 77
GenBank mRNA AB445101.1 - 3 - 4 418 77
AF015608.1 - 3 - 4 432 77
AF247662.1 - 1 - 2 187 77
AF274003.1 - 3 - 4 394 77
AK289955.1 - 5 - 6 653 77
AK303100.1 - 3 - 4 366 77
AK316132.1 - 3 - 4 695 77
AL520572.3 - 4 - 5 676 77
AL559988.3 - 3 - 4 480 77
BC001659.2 - 4 - 5 583 2959954 77
BC001838.2 - 4 - 5 583 2959954 77
BC108316.1 - 4 - 5 704 6151864 77
BQ059026.1 - 2 561 77
DA728900.1 - 4 - 5 595 77
L37368.1 - 4 - 5 967 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RNPS1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
regulation of alternative mRNA splicing, via spliceosome
mRNA splicing, via spliceosome
transcription, DNA-templated
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
positive regulation of apoptotic process
negative regulation of mRNA splicing, via spliceosome
nucleotide binding
RNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0