This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00992170_m1
See other GRK5 GE Assays ›
Gene Symbol:
GRK5
Gene Name:
G protein-coupled receptor kinase 5
Gene Aliases:
GPRK5
Chromosome Location:
Chr.10: 119207685 - 119455619 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00992170_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRK5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005308.2 NP_005299.1 10 - 11 1197 74
XM_005269708.1 XP_005269765.1 9 - 10 1087 74
GenBank mRNA AK301450.1 - 9 - 10 888 74
AK313012.1 - 10 - 11 1044 74
BC064506.1 - 10 - 11 1191 6482467 74
L15388.1 - 10 - 11 1184 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009067.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000865.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GRK5

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
apoptotic process
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
tachykinin receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of cell proliferation
Wnt signaling pathway
response to drug
negative regulation of apoptotic process
fat cell differentiation
protein autophosphorylation
regulation of cell cycle
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
protein kinase C binding
protein binding
ATP binding
phospholipid binding
beta-adrenergic receptor kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10010463.hg.1 ENST00000369108.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010463.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10010463.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10010463.hg.1 TC10000865.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000369108.4 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay TC1000009067.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay PSR1000162098.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000162098.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay PSR1000162098.hg.1 ENST00000392870.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000162098.hg.1 NM_005308.2 Refseq
Affymetrix Human Clariom D Assay PSR1000162098.hg.1 TC1000009067.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0