This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00991337_m1
See other MYO15A GE Assays ›
Gene Symbol:
MYO15A
Gene Name:
myosin XVA
Gene Aliases:
DFNB3, MYO15
Chromosome Location:
Chr.17: 18108706 - 18180227 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00991337_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYO15A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016239.3 NP_057323.3 42 - 43 8300 80
XM_017024714.1 XP_016880203.1 39 - 40 8600 80
XM_017024715.1 XP_016880204.1 40 - 41 8663 80
GenBank mRNA AF144094.1 - 42 - 43 8300 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700007100.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000205.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MYO15A

Panther Classification:

Molecular Function -

actin binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
locomotory behavior
inner ear morphogenesis
motor activity
actin binding
calmodulin binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17002929.hg.1 ENST00000205890.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17002929.hg.1 ENST00000418233.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17002929.hg.1 NM_016239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17002929.hg.1 TC17000205.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17002930.hg.1 ENST00000205890.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17002930.hg.1 NM_016239.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17002930.hg.1 TC17000205.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 ENST00000205890.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 ENST00000418233.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 ENST00000615845.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 NM_016239.3 Refseq
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 ENST00000205890.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 ENST00000418233.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 ENST00000615845.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 NM_016239.3 Refseq
Affymetrix Human Clariom D Assay PSR1700150131.hg.1 TC1700007100.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0