This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00991064_m1
See other S100Z GE Assays ›
Gene Symbol:
S100Z
Gene Name:
S100 calcium binding protein Z
Gene Aliases:
Gm625, S100-zeta
Chromosome Location:
Chr.5: 76849953 - 76952882 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00991064_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100Z
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130772.3 NP_570128.2 2 - 3 272 65
XM_011543239.2 XP_011541541.1 2 - 3 320 65
XM_011543240.2 XP_011541542.1 2 - 3 320 65
XM_011543241.2 XP_011541543.1 2 - 3 320 65
XM_011543244.2 XP_011541546.1 2 - 3 287 65
XM_017009171.1 XP_016864660.1 2 - 3 512 65
XM_017009172.1 XP_016864661.1 2 - 3 523 65
GenBank mRNA BC022320.1 - 2 - 3 273 4696667 65
BX281742.1 - 2 - 3 174 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500007836.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000373.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC022320.1,NM_130772.3,XM_011543239.2,XM_011543240.2,XM_011543241.2,XM_011543244.2,XM_017009171.1,XM_017009172.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100Z

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05005234.hg.1 ENST00000317593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005234.hg.1 ENST00000513010.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05005234.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05005234.hg.1 TC05000373.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153624.hg.1 TC0500007836.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay TC0500007836.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 ENST00000317593.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 ENST00000513010.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 ENST00000613039.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 NM_130772.3 Refseq
Affymetrix Human Clariom D Assay PSR0500153623.hg.1 TC0500007836.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0