Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030112.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030112.hg.1 |
ENST00000366592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030112.hg.1 |
ENST00000391852.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030112.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030112.hg.1 |
TC01001950.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
ENST00000477559.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012089.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000366591.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000366592.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000419162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
ENST00000454895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
NM_003272.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100188958.hg.1 |
TC0100012089.hg.1 |
Clariom D TC |