This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00990710_m1
See other GPR137B GE Assays ›
Gene Symbol:
GPR137B
Gene Name:
G protein-coupled receptor 137B
Gene Aliases:
TM7SF1
Chromosome Location:
Chr.1: 236142502 - 236211059 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00990710_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR137B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003272.3 NP_003263.1 2 - 3 558 100
XM_017002209.1 XP_016857698.1 2 - 3 588 100
XM_017002210.1 XP_016857699.1 2 - 3 588 100
GenBank mRNA AF027826.1 - 2 - 3 518 100
AK223632.1 - 2 - 3 537 100
AK295408.1 - 3 - 4 470 100
AK301552.1 - 3 - 4 490 100
AK312908.1 - 2 - 3 551 100
AY242135.1 - 2 - 3 467 100
BC012100.1 - 2 - 3 558 4643216 100
CR541770.1 - 2 - 3 467 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100012089.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001950.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR137B

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01030111.hg.1 ENST00000366592.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01030111.hg.1 ENST00000391852.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01030111.hg.1 NM_003272.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01030111.hg.1 TC01001950.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay PSR0100188956.hg.1 TC0100012089.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366591.4 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000366592.7 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000419162.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000454895.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 ENST00000477559.2 Ensembl
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq
Affymetrix Human Clariom D Assay TC0100012089.hg.1 NM_003272.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0