This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00989713_m1
See other CFI GE Assays ›
Gene Symbol:
CFI
Gene Name:
complement factor I
Gene Aliases:
AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF
Chromosome Location:
Chr.4: 109731221 - 109802225 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00989713_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CFI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005262976.1 XP_005263033.1 7 - 8 1169 136
XM_017008165.1 XP_016863654.1 7 - 8 1169 136

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
receptor-mediated endocytosis
complement activation, classical pathway
regulation of complement activation
innate immune response
serine-type endopeptidase activity
scavenger receptor activity
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header