This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00984811_m1
See other USB1 GE Assays ›
Gene Symbol:
USB1
Gene Name:
U6 snRNA biogenesis phosphodiesterase 1
Gene Aliases:
C16orf57, HVSL1, Mpn1, PN, hUsb1
Chromosome Location:
Chr.16: 57999600 - 58021623 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00984811_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: USB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024598.3 NP_078874.2 4 - 5 613 87
XM_005256144.3 XP_005256201.1 4 - 5 488 87
XM_011523328.2 XP_011521630.1 4 - 5 589 87
XM_011523329.2 XP_011521631.1 4 - 5 471 87
GenBank mRNA AK023216.1 - 4 - 5 585 87
AK293220.1 - 4 - 5 423 87
AK298473.1 - 5 - 6 460 87
BC004415.1 - 4 - 5 549 3641317 87
BC006291.2 - 4 - 5 539 4025993 87
BC007774.2 - 4 - 5 543 4302289 87
BC021554.1 - 4 - 5 533 4541229 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008020.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000499.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene USB1

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
U6 snRNA 3'-end processing
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000219281.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000423271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000539737.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000561743.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000562534.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000562534.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000562534.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000562534.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000562534.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000562534.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000562534.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563149.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000563207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000564387.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000564387.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000564387.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000564387.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000564387.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000564387.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000565662.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000566292.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000568848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000569252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000569252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000569252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000569252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000569252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000569252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000569252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000624720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000624720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000624720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000624720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000624720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000624720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 ENST00000624720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001195302.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_001204911.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008020.hg.1 NM_024598.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0