This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00984358_g1
See other SLC6A4 GE Assays ›
Gene Symbol:
SLC6A4
Gene Name:
solute carrier family 6 member 4
Gene Aliases:
5-HTT, 5-HTTLPR, 5HTT, HTT, OCD1, SERT, SERT1, hSERT
Chromosome Location:
Chr.17: 30194319 - 30235968 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00984358_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC6A4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045.5 NP_001036.1 1548 63
GenBank mRNA AY902473.1 - 5 - 6 1043 63
BC069484.1 - 5 - 6 1020 7262380 63
L05568.1 - 5 - 6 1044 63
X70697.1 - 5 - 6 1082 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700010272.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001326.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC6A4

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
serotonin transport
chemical synaptic transmission
response to nutrient
memory
circadian rhythm
response to toxic substance
positive regulation of gene expression
monoamine transport
thalamus development
negative regulation of cerebellar granule cell precursor proliferation
negative regulation of synaptic transmission, dopaminergic
response to estradiol
social behavior
vasoconstriction
response to drug
sperm ejaculation
negative regulation of neuron differentiation
positive regulation of cell cycle
negative regulation of organ growth
brain morphogenesis
protein oligomerization
protein homooligomerization
serotonin uptake
transmembrane transport
cellular response to retinoic acid
cellular response to cGMP
serotonin:sodium symporter activity
protein binding
monoamine transmembrane transporter activity
serotonin transmembrane transporter activity
myosin binding
syntaxin-1 binding
Rab GTPase binding
cocaine binding
protein homodimerization activity
metal ion binding
nitric-oxide synthase binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000578609.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000578609.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000578609.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17017753.hg.1 ENST00000261707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17017753.hg.1 ENST00000394821.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17017753.hg.1 ENST00000401766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17017753.hg.1 NM_001045.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17017753.hg.1 TC17001326.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000578609.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000578609.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000578609.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 ENST00000579221.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay TC1700010272.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 ENST00000261707.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 ENST00000394821.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 ENST00000401766.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 NM_001045.5 Refseq
Affymetrix Human Clariom D Assay PSR1700180810.hg.1 TC1700010272.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm