This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00984191_m1
See other CATSPERB GE Assays ›
Gene Symbol:
CATSPERB
Gene Name:
cation channel sperm associated auxiliary subunit beta
Gene Aliases:
C14orf161, CatSper(beta)
Chromosome Location:
Chr.14: 91580774 - 91732086 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00984191_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CATSPERB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024764.3 NP_079040.2 20 - 21 2562 93
GenBank mRNA AK024360.1 - 6 - 7 1090 93
AK126034.1 - 20 - 21 2546 93
BC126214.1 - 20 - 21 2516 8991930 93
BC126216.1 - 20 - 21 2516 8991932 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400010776.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001439.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CATSPERB

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s)

multicellular organism development
response to progesterone
sperm-egg recognition
sperm capacitation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000556429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000556429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000556429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14016069.hg.1 ENST00000256343.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14016069.hg.1 NM_024764.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14016069.hg.1 TC14001439.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000553329.5 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000556429.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000556429.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000556429.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000556901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay TC1400010776.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay PSR1400183066.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400183066.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400183066.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay PSR1400183066.hg.1 ENST00000256343.7 Ensembl
Affymetrix Human Clariom D Assay PSR1400183066.hg.1 ENST00000557036.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400183066.hg.1 NM_024764.3 Refseq
Affymetrix Human Clariom D Assay PSR1400183066.hg.1 TC1400010776.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0