This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00983587_g1
See other CFAP73 GE Assays ›
Gene Symbol:
CFAP73
Gene Name:
cilia and flagella associated protein 73
Gene Aliases:
CCDC42B, MIA2
Chromosome Location:
Chr.12: 113149162 - 113159276 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00983587_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CFAP73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144872.1 NP_001138344.1 3 - 4 266 62
XM_011538327.2 XP_011536629.1 3 - 4 962 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200008929.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000888.hg.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12011739.hg.1 ENST00000335621.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12011739.hg.1 NM_001144872.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12011739.hg.1 TC12000888.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay TC1200008929.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 ENST00000335621.10 Ensembl
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 ENST00000550918.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 ENST00000551256.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 NM_001144872.2 Refseq
Affymetrix Human Clariom D Assay PSR1200166379.hg.1 TC1200008929.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0