This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00981374_m1
See other POLQ GE Assays ›
Gene Symbol:
POLQ
Gene Name:
DNA polymerase theta
Gene Aliases:
PRO0327
Chromosome Location:
Chr.3: 121431420 - 121546006 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00981374_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLQ
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199420.3 NP_955452.3 17 - 18 5896 95
XM_011512343.2 XP_011510645.1 17 - 18 5884 95
XM_011512347.2 XP_011510649.1 17 - 18 5884 95
XM_011512348.2 XP_011510650.1 16 - 17 5753 95
XM_011512352.2 XP_011510654.1 17 - 18 5884 95
XM_011512354.2 XP_011510656.1 5 - 6 3642 95
XM_017005565.1 XP_016861054.1 11 - 12 4748 95
GenBank mRNA AB209867.1 - 17 - 18 5823 95
AF043628.1 - 2 - 3 1466 95
AF052573.1 - 2 - 3 3696 95
AY032677.1 - 18 - 19 6169 95
AY338826.2 - 17 - 18 5896 95
CR936627.1 - 16 - 17 7946 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012164.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001695.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLQ

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA-dependent DNA replication
DNA repair
base-excision repair
double-strand break repair
cellular response to DNA damage stimulus
somatic hypermutation of immunoglobulin genes
protein homooligomerization
DNA biosynthetic process
double-strand break repair via alternative nonhomologous end joining
negative regulation of double-strand break repair via homologous recombination
chromatin binding
damaged DNA binding
DNA-directed DNA polymerase activity
protein binding
ATP binding
single-stranded DNA-dependent ATPase activity
5'-deoxyribose-5-phosphate lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03030618.hg.1 ENST00000264233.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030618.hg.1 ENST00000393672.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030618.hg.1 ENST00000543272.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03030618.hg.1 NM_199420.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03030618.hg.1 TC03001695.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300190536.hg.1 ENST00000264233.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190536.hg.1 ENST00000621776.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300190536.hg.1 NM_199420.3 Refseq
Affymetrix Human Clariom D Assay PSR0300190536.hg.1 ENST00000264233.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300190536.hg.1 ENST00000621776.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300190536.hg.1 NM_199420.3 Refseq
Affymetrix Human Clariom D Assay PSR0300190536.hg.1 TC0300012164.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0