Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056326.hg.1 |
ENST00000367350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056326.hg.1 |
NM_014875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056326.hg.1 |
TC01003679.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
ENST00000367350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
ENST00000614960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
NM_001305792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
NM_014875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
ENST00000367350.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
ENST00000614960.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
NM_001305792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
NM_014875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227701.hg.1 |
TC0100016891.hg.1 |
Clariom D TC |