This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00978188_m1
See other GPR160 GE Assays ›
Gene Symbol:
GPR160
Gene Name:
G protein-coupled receptor 160
Gene Aliases:
GPCR1, GPCR150
Chromosome Location:
Chr.3: 170037323 - 170085415 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00978188_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR160
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014373.2 NP_055188.1 3 - 4 523 154
XM_005247346.3 XP_005247403.1 4 - 5 1133 154
XM_005247347.3 XP_005247404.1 2 - 3 169 154
XM_006713582.3 XP_006713645.1 3 - 4 485 154
XM_011512678.2 XP_011510980.1 4 - 5 815 154
XM_017006161.1 XP_016861650.1 4 - 5 626 154
XM_017006163.1 XP_016861652.1 3 - 4 1147 154
GenBank mRNA AJ249248.1 - 2 - 3 254 154
AK311293.1 - 4 - 5 1133 154
BC000181.2 - 3 - 4 458 3048193 154
BX366931.2 - 3 - 4 523 155

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300009459.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000910.hg.1

Additional Information:

This assay is in the 5'UTR region of the AJ249248.1,BC000181.2,NM_014373.2,XM_005247346.3,XM_005247347.3,XM_006713582.3,XM_011512678.2,XM_017006161.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR160

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03015931.hg.1 ENST00000355897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03015931.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03015931.hg.1 TC03000910.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03015933.hg.1 ENST00000355897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03015933.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03015933.hg.1 TC03000910.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03015932.hg.1 ENST00000355897.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03015932.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03015932.hg.1 TC03000910.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay PSR0300167974.hg.1 TC0300009459.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay PSR0300167976.hg.1 TC0300009459.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay PSR0300167975.hg.1 TC0300009459.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0