This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00978184_m1
See other GPR160 GE Assays ›
Gene Symbol:
GPR160
Gene Name:
G protein-coupled receptor 160
Gene Aliases:
GPCR1, GPCR150
Chromosome Location:
Chr.3: 170037323 - 170085415 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00978184_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR160
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017006162.1 XP_016861651.1 3 - 4 476 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017006162.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000355897.9 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000461049.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000473675.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000476668.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000478364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482710.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000482813.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000485735.6 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000491740.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492492.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 ENST00000492924.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq
Affymetrix Human Clariom D Assay TC0300009459.hg.1 NM_014373.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header